Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UNKL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567317
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567317 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567317 Supplier Invitrogen™ Supplier No. PA567317
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRRQLDEAKRK IRQWEESWQQ VKQVCDAWQR EAQEAKERAR VADSDRQLAL QKKEEVEAQV KQ Highest antigen sequence identity to the following orthologs - mouse 95%, rat 97%.

May participate in a protein complex showing an E3 ligase activity regulated by RAC1. Ubiquitination is directed towards itself and possibly other substrates, such as SMARCD2/BAF60b. Intrinsic E3 ligase activity has not been proven.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UNKL
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UNKL
Gene Accession No. Q9H9P5
Gene Alias 1300004G08Rik; C16orf28; Grailexp predicted; putative E3 ubiquitin-protein ligase UNKL; RGD1565247; RING finger protein unkempt-like; RING finger protein unkempt-like; putative E3 ubiquitin-protein ligase UNKL; si:ch211-66h3.3; unk like zinc finger; unkempt family like zinc finger; unkempt family zinc finger-like; unkempt homolog (Drosophila)-like; unkempt homolog-like; unkempt-like; unkempt-like (Drosophila); Unkl; ZC3H5L; ZC3HDC5L; Zinc finger CCCH domain-containing protein 5-like
Gene Symbols UNKL
Host Species Rabbit
Immunogen Recombinant Human UNKL. Recombinant protein control fragment (Product #RP-107959).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64718
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.