Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UNKL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA569227
Description
This target displays homology in the following species: Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
May participate in a protein complex showing an E3 ligase activity regulated by RAC1. Ubiquitination is directed towards itself and possibly other substrates, such as SMARCD2/BAF60b. Intrinsic E3 ligase activity has not been proven.
Specifications
UNKL | |
Polyclonal | |
Unconjugated | |
UNKL | |
1300004G08Rik; C16orf28; Grailexp predicted; putative E3 ubiquitin-protein ligase UNKL; RGD1565247; RING finger protein unkempt-like; RING finger protein unkempt-like; putative E3 ubiquitin-protein ligase UNKL; si:ch211-66h3.3; unk like zinc finger; unkempt family like zinc finger; unkempt family zinc finger-like; unkempt homolog (Drosophila)-like; unkempt homolog-like; unkempt-like; unkempt-like (Drosophila); Unkl; ZC3H5L; ZC3HDC5L; Zinc finger CCCH domain-containing protein 5-like | |
Rabbit | |
Affinity chromatography | |
RUO | |
74154 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5FWH2 | |
UNKL | |
synthetic peptide directed towards the middle region of mouse UNKL. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction