Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
uNOS Antibody (NOS-3F7-B11 B5), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202801
Description
uNOS Monoclonal specifically detects uNOS in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
uNOS | |
Monoclonal | |
Unconjugated | |
bNOS, Constitutive NOS, IHPS 1, IHPS1, N NOS, NC NOS, Neuronal nitric oxide synthase, Neuronal NOS, Nitric oxide synthase 1, Nitric oxide synthase 1 neuronal, Nitric oxide synthase brain, nNOS, NOS, Nos 1, NOS type I, NOS1 | |
Mouse | |
Unpurified | |
RUO | |
Primary | |
Detects brain Nitric Oxide Synthase (bNOS), inducible NOS (iNOS) and epithelial NOS (eNOS). | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgM |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
NOS-3F7-B11 B5 | |
Western Blot 1:50 - 1:500, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:100 | |
NOS2 | |
Purified bovine bNOS. | |
200 μL | |
Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission | |
4843 | |
Human, Mouse, Rat, Bovine | |
Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction