Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UNR Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501564418
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-4418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-4418 Supplier Bethyl Laboratories, Inc Supplier No. A303160A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Guinea pig_10141

CSDE1 gene ontology annotations related to this gene include regulation of transcription, DNA-templated.

Specifications

Antigen UNR
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CSDE1
Gene Accession No. O75534, Q91W50
Gene Alias D1S155E, RP5-1000E10.3, UNR, N-ras upstream gene protein, NRAS-related, cold shock domain-containing protein E1, upstream of NRAS
Gene Symbols CSDE1
Host Species Rabbit
Immunogen Between 748 and 798
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 229663, 7812
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.