Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
uPA (PLAU) Mouse anti-Human, PE, Clone: UK98/6 (also known as UK 98/6; 98/6), BD Phrmingen™
Mouse Monoclonal Antibody
$289.50
Specifications
| Antigen | uPA (PLAU) |
|---|---|
| Clone | UK98/6 (also known as UK 98/6; 98/6) |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
uPA (PLAU) Monoclonal antibody specifically detects uPA (PLAU) in Human samples. It is validated for Flow cytometrySpecifications
| uPA (PLAU) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 5328 | |
| IgG1 κ | |
| Affinity Chromatography |
| UK98/6 (also known as UK 98/6; 98/6) | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human | |
| ATF; PLAU; U-plasminogen activator; UPA; u-PA; urokinase-type plasminogen activator | |
| Purified Human Urokinase-type Plasminogen Activator | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title