Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
uPAR Antibody (014), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290454AF700
Description
uPAR Monoclonal specifically detects uPAR in Mouse samples. It is validated for ELISA.Specifications
uPAR | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
14 | |
ELISA | |
CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse uPAR (NP_035243.1; Met1-Thr297). | |
0.1 mL | |
Cancer | |
5329 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction