Learn More
uPAR Antibody (109801), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB531MFV610
Description
uPAR Monoclonal antibody specifically detects uPAR in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blocking Assay, CyTOF-reportedSpecifications
| uPAR | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cancer | |
| 5329 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Blocking Assay, CyTOF | |
| 109801 | |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-reported | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse uPAR isoform 1, Leu24-Thr297, Accession # P35456 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a |