Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
uPAR Antibody (IIIB11), Novus Biologicals™

Mouse Monoclonal Antibody
$243.50 - $697.00
Specifications
Antigen | uPAR |
---|---|
Clone | IIIB11 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
uPAR Monoclonal specifically detects uPAR in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
uPAR | |
Monoclonal | |
Purified | |
Cancer | |
PBS with 0.1% BSA with 0.02% Sodium Azide | |
5329 | |
uPAR (D2 domain) | |
Primary |
IIIB11 | |
Unconjugated | |
Mouse | |
Human | |
CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
PLAUR | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title