Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

uPAR (CD87), Mouse anti-Hamster, Clone: 5D6, Millipore Sigma™

Catalog No. MABC109925 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC109925 25 μg
MABC1099100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC109925 Supplier MilliporeSigma Supplier No. MABC109925UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-uPAR (CD87), clone 5D6, Cat. No. MABC1099, is a mouse monoclonal antibody that detects Urokinase plasminogen activator surface receptor and has been tested for use in Flow Cytometry, Immunocytochemistry, and Western Blotting.

Urokinase plasminogen activator surface receptor (UniProt: Q9R119; also known as uPAR) is encoded by the Upar gene (Gene ID: 100689422) in Chinese hamster. uPAR is a GPI-anchored cell membrane receptor for urokinase plasminogen activator (uPA), an enzyme responsible for the activation of plasmin and degradation of extracellular matrix components. Higher expression of uPAR has been reported in many human cancers, including non-small cell lung cancer and colorectal cancer. Its higher levels correlate with poor prognosis and early invasion and metastasis. uPAR is synthesized with a signal peptide (aa 1-18), which is subsequently cleaved off to generate the mature form, which contains three homologus domains (DI, DII, and DIII). uPAR is also reported to serve as an adhesion receptor for vitronectin and a direct interaction between uPAR and vitronectin is shown to be important for inducing changes in cell morphology, cell migration, and signaling. Blocking of uPAR activity can impair cell adhesion and migration in RAS mutated cancer cells. (Ref.: Di Mauro, C et al. (2017). Scientific Reports 7, Article number: 9388).
TRUSTED_SUSTAINABILITY

Specifications

Antigen uPAR (CD87)
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 5D6
Conjugate Unconjugated
Dilution Western Blotting Analysis: A representative lot detected uPAR (CD87) in Western Blotting applications (Maeda, Y., et. al. (2007). Mol Biol Cell. 18(4):1497-506; Lee, G.H., et. al. (2016). J Cell Biol. 215(5):705-718; Hirata, T., et. al. (2018). Nat Commun. 9(1):405).Flow Cytometry Analysis: A representative lot detected uPAR (CD87) in Flow Cytometry applications (Ashida, H., et. al. (2006). J Biol Chem. 281(2):896-904; Lee, G.H., et. al. (2016). J Cell Biol. 215(5):705-718; Hirata, T., et. al. (2018). Nat Commun. 9(1):405; Hirata, T., et. al. (2013). J Biochem. 154(3):257-64).Immunocytochemistry Analysis: A representative lot detected uPAR (CD87) in Immunocytochemistry applications (Kanzawa, N., et. al. (2012). J Lipid Res. 53(4):653-63).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Urokinase plasminogen activator surface receptor;CD87
Gene Symbols Upar
Host Species Mouse
Immunogen Raft fraction of the plasma membrane from Chinese hamster ovary (CHO) cells.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis & Cancer
Primary or Secondary Primary
Gene ID (Entrez) XP_007647182.1
Test Specificity Clone 5D6 detects urokinase plasminogen activator surface receptor. It targets an epitope with in the extracellular domain.
Target Species Hamster
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.