Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ uPAR (CD87) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579826
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579826 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579826 Supplier Invitrogen™ Supplier No. PA579826
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat pancreas tissue, rat small intestine tissue, rat kidney tissue, mouse brain tissue, mouse pancreas tissue, mouse kidney tissue, mouse testis tissue.

The urokinase-type plasminogen activator receptor is a key molecule in the regulation of cell-surface plasminogen activation and plays an important role in many normal as well as pathologic processes. The human PLAUR cDNA encodes 335 amino acids including a predicted signal peptide of 22 residues and a hydrophobic C-terminal portion. It produces a highly glycosylated protein of about 50 kD in monocytes where it is anchored to the plasma membrane by glycosyl-phosphatidylinositol linkage. PLAUR, also known as UPAR, is directly associated with the carbohydrate-binding domain of SELL in the membrane of neutrophils, an association analogous to that between PLAUR and beta-2 integrins. PLAUR-mediated calcium mobilization is SELL dependent. UPAR mRNA levels correlate with the invasive potential of endometrial carcinomas and show a 33-fold increase in UPAR mRNA levels in advanced clinical stage endometrial tumors compared with normal endometrial tissue. Furthermore, the increase in UPAR mRNA levels correlated linearly with the progression of disease stage. UPAR protein expressioin correlated positively with rate of recurrence and mortality in patients with endometrial cancer. UPAR appears to be a useful prognostic marker for advanced endometrial cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen uPAR (CD87)
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene PLAUR
Gene Accession No. P35456, P49616
Gene Alias Cd87; MO3; Monocyte activation antigen Mo3; Par; plasminogen activator urokinase receptor 3; plasminogen activator, urokinase receptor; PLAUR; Plaur3; sCD87; soluble CD87; uPAR; U-PAR; uPAR-2; uPAR-3; u-plasminogen activator receptor form 2; urinary plasminogen activator receptor 2; urinary plasminogen activator receptor 3; URKR; urokinase plasminogen activator receptor; urokinase plasminogen activator surface receptor; urokinase-type plasminogen activator (uPA) receptor; urokinase-type plasminogen activator receptor
Gene Symbols PLAUR
Host Species Rabbit
Immunogen E. coli-derived rat uPA Receptor recombinant protein (Position: L25-R261).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18793, 50692
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.