Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Idex Bottle Caps

Click to view available options
For Use With (Equipment):
0.06 in. O.D. tubing
Thread Size (Metric):
GL 38
Color:
Aqua Blue
Specifications
Specifications
Color | Aqua Blue |
Material | Polypropylene |
No. per Case | 1 Cs. |
Thread Size (Metric) | GL 38 |
Product Type | Cap |
For Use With (Equipment) | 0.06 in. O.D. tubing |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction