Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Idex Stainless-Steel Tubing 0.020 in.OD 0.005 in.ID

Catalog No. 05701135
Encompass
Encompass_Preferred
Change view
Click to view available options
Overall Length (Metric):
10 cm
20 cm
1.5 m
Diameter (Metric) Inner:
0.5 mm
1.0 mm
Material:
Stainless Steel (304)
Stainless Steel (316)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Overall Length (Metric) Diameter (Metric) Inner Material
05-701-135 1.5 m 1.0 mm Stainless Steel (316)
05-701-130 10 cm 1.0 mm Stainless Steel (316)
05-701-96 20 cm 0.5 mm Stainless Steel (304)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 05-701-135 Supplier Idex Supplier No. U144
Add to Cart
Add to Cart

Seamless, precut stainless steel tubing is specially prepared for the exacting requirements of todays analyses

Stainless steel is naturally self-passivating, forming an oxidation layer on newly created surfaces. Idex Heath & Science Scientific takes extra steps to ensure the chemical resistance of the stainless steel tubing by manually passivating before and after the tubing is cut into specified lengths.

  • Precut 316 stainless steel tubing:
  • Ends are machine cut and polished
  • Edges are deburred inside and outside
  • Tubing is passivated
  • Inside is flushed with reagent-grade isopropanol
  • Color-coded banding for easy identification of the inner diameter
  • All stainless-steel tubing longer than 1m is coiled

Specifications

Type Tubing
Diameter (English) Inner 1/16 in.
Diameter (English) Outer 0.040 in.
Diameter (Metric) Inner 1.0 mm
Overall Length (English) 5 ft.
Overall Length (Metric) 1.5 m
Material Stainless Steel (316)
Maximum Pressure 8100 psi (558 bar)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.