Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UPF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541262 Supplier Invitrogen™ Supplier No. PA541262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: RGTPKGKTGR GGRQKNRFGL PGPSQTNLPN SQASQDVASQ PFSQGALTQG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%; Zebrafish: 83%.

Upf1 was identified as an active component in nonsense-mediated decay (NMD), an mRNA surveillance mechanism in eukaryotic cells that degrades mRNAs containing premature termination codons. Upf1 was found to be an ATP-dependent RNA helicase in the cytoplasm and was later shown to be a component of cytoplasmic P-bodies. Upf1 phosphorylation mediates the repression of translation that accompanies NMD, allowing mRNA accessibility to the NMD machinery. Two other active components of NMD, Upf2 and Upf3, were also identified and described as having perinuclear and nucleocytoplasmic localization, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPF1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UPF1
Gene Accession No. Q92900
Gene Alias ATP-dependent helicase RENT1; B430202H16Rik; delta helicase; FLJ43809; FLJ46894; HUPF1; KIAA0221; LOW QUALITY PROTEIN: regulator of nonsense transcripts 1; mUpf1; Nonsense mRNA reducing factor 1; NORF1; pNORF1; PNORF-1; Regulator of nonsense transcripts 1; Regulator of nonsense transcripts 1 (Nonsense mRNA reducing factor 1) (NORF1) (Up-frameshift suppressor 1 homolog); Rent1; smg-2; smg-2 homolog, nonsense mediated mRNA decay factor; Upf1; UPF1 regulator of nonsense transcripts homolog; UPF1 regulator of nonsense transcripts homolog (yeast); UPF1 RNA helicase and ATPase; UPF1, RNA helicase and ATPase; Upflp; up-frameshift mutation 1 homolog; up-frameshift suppressor 1 homolog; wu:fi40f07; wu:fj48a01; yeast Upf1p homolog; zgc:55472
Gene Symbols UPF1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human UPF1.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5976
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.