Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UPF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566888
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566888 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566888 Supplier Thermo Scientific Supplier No. PA566888
Add to Cart
Add to Cart

UPF2 Polyclonal Antibody

UPF2 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen UPF2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UPF2
Gene Accession No. Q9HAU5
Gene Alias HUPF2; KIAA1408; Nonsense mRNA reducing factor 2; regulator of nonsense transcripts 2; RENT2; smg-3; smg-3 homolog, nonsense mediated mRNA decay factor; UPF2; UPF2 regulator of nonsense transcripts homolog (yeast); up-frameshift suppressor 2 homolog; yeast Upf2p homolog
Gene Symbols UPF2
Host Species Rabbit
Immunogen Recombinant Human UPF2. Recombinant protein control fragment (Product #RP-107535).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26019
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.