Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UPF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577128
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located in the perinuclear area. It interacts with translation release factors and the proteins that are functional homologs of yeast Upf1p and Upf3p. Two splice variants have been found for this gene; both variants encode the same protein.
Specifications
UPF2 | |
Polyclonal | |
Unconjugated | |
UPF2 | |
HUPF2; KIAA1408; Nonsense mRNA reducing factor 2; regulator of nonsense transcripts 2; RENT2; smg-3; smg-3 homolog, nonsense mediated mRNA decay factor; UPF2; UPF2 regulator of nonsense transcripts homolog (yeast); up-frameshift suppressor 2 homolog; yeast Upf2p homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26019, 361271 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HAU5 | |
UPF2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1153-1272 of human UPF2. | |
100 μL | |
Primary | |
Human, Rat, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction