Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UPF2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA528629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528629 Supplier Thermo Scientific Supplier No. PA528629

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located in the perinuclear area. It interacts with translation release factors and the proteins that are functional homologs of yeast Upf1p and Upf3p. Two splice variants have been found for this gene; both variants encode the same protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPF2
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene UPF2
Gene Accession No. Q9HAU5
Gene Alias HUPF2; KIAA1408; Nonsense mRNA reducing factor 2; regulator of nonsense transcripts 2; RENT2; smg-3; smg-3 homolog, nonsense mediated mRNA decay factor; UPF2; UPF2 regulator of nonsense transcripts homolog (yeast); up-frameshift suppressor 2 homolog; yeast Upf2p homolog
Gene Symbols UPF2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to a region within amino acids 1200 and 1272 of Human Rent2
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26019, 326622
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.