Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UPF3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584488
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584488 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584488 Supplier Invitrogen™ Supplier No. PA584488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FLETYCVEEE KTSANPETLL GEMEAKTREL IARRTTPLLE YIKNRKLEKQ R Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 88%.

UPF3A is part of a multiprotein post-splicing mRNP complex involved in both mRNA nuclear export and mRNA surveillance. UPF3A is involved in nonsense-mediated decay (NMD) of mRNAs containing premature stop codons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPF3A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UPF3A
Gene Accession No. Q9H1J1
Gene Alias 2600001C03Rik; 4930546M19Rik; hUpf3; HUPF3A; nonsense mRNA reducing factor 3A; Regulator of nonsense transcripts 3A; RENT3A; UPF3; UPF3 regulator of nonsense transcripts homolog A (yeast); UPF3A; Up-frameshift suppressor 3 homolog A
Gene Symbols UPF3A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UPF3A. Recombinant protein control fragment (Product #RP-107052).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65110
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.