Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UPF3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578562
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578562 Supplier Invitrogen™ Supplier No. PA578562
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Rhesus Monkey (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

UPF3B is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The protein is one of two functional homologs to yeast Upf3p. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. The encoded protein is one of two functional homologs to yeast Upf3p. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein binds to the mRNA and remains bound after nuclear export, acting as a nucleocytoplasmic shuttling protein. It forms with Y14 a complex that binds specifically 20 nt upstream of exon-exon junctions. This gene is located on the long arm of chromosome X. Two splice variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPF3B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene UPF3B
Gene Accession No. Q9BZI7
Gene Alias 5730594O13Rik; AI317193; AW541158; HUPF3B; hUpf3p-X; mental retardation, X-linked 62; MRX62; MRXS14; Nonsense mRNA reducing factor 3B; regulator of nonsense transcripts 3B; RENT3B; RGD1560264; Unknown (protein for MGC:134301); UPF3 regulator of nonsense transcripts homolog B; UPF3 regulator of nonsense transcripts homolog B (yeast); UPF3 regulator of nonsense transcripts-like protein B; UPF3B; UPF3B pseudogene 1; UPF3B pseudogene 2; UPF3B pseudogene 3; UPF3B regulator of nonsense mediated mRNA decay; UPF3BP1; UPF3BP2; UPF3BP3; Upf3p-X; UPF3X; up-frameshift suppressor 3 homolog B; Up-frameshift suppressor 3 homolog on chromosome X
Gene Symbols UPF3B
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human UPF3B. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65109
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.