Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UPK1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572711
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572711 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572711 Supplier Invitrogen™ Supplier No. PA572711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UPK1B Polyclonal Antibody for Western Blot, IHC (P), ELISA

Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in normal bladder epithelial physiology, possibly in regulating membrane permeability of superficial umbrella cells or in stabilizing the apical membrane through AUM/cytoskeletal interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPK1B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene UPK1B
Gene Accession No. O75841, Q566D0, Q9Z2C6
Gene Alias AI413235; tetraspan; tetraspanin-20; Tspan20; Tspan-20; UP1b; UPIb; Upk1; Upk1b; uroplakin 1B; uroplakin Ib; Uroplakin-1b
Gene Symbols UPK1B
Host Species Rabbit
Immunogen UPK1B antibody for a 17 amino acid synthetic peptide near the center of human UPK1B. The immunogen is within amino acids 110 - 160 of UPK1B.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22268, 303924, 7348
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.