Learn More
Invitrogen™ UPK3A Monoclonal Antibody (SP73)

Description
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is bladder carcinoma.
Specifications
Specifications
| Antigen | UPK3A |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP73 |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.1% sodium azide; pH 7.6 |
| Gene | Upk3a |
| Gene Accession No. | O75631 |
| Gene Alias | 1110017C07Rik; proplakin IIIa; UP3a; UPIII; UPIIIA; Upk3; UPK3A; uroplakin 3; uroplakin 3A; uroplakin III; Uroplakin-3a |
| Gene Symbols | Upk3a |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.