Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UPK3A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587581 Supplier Invitrogen™ Supplier No. PA587581
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNLQPQLASV TFATNNPTLT TVALEKPLCM FDSKEALTGT HEVYLYVLVD SAISRNASVQ DSTNTPLGST FLQTEGGRTG PYKAVAFDLI PCSDLPSLDA IGDVSKASQI LNAYLVRVGA NGTCLWDPNF QGLCNAPLSA ATEYRFKYVL VNMSTGLVED QTLWSDPIRT NQLTPYSTID TWPGRRSGG; Positive Samples: HeLa, HL-60, Mouse brain, Mouse kidney, Rat kidney; Cellular Location: Endoplasmic reticulum membrane, Single-pass type I membrane protein.

This gene encodes a member of the uroplakin family, a group of transmembrane proteins that form complexes on the apical surface of the bladder epithelium. Mutations in this gene may be associated with renal adysplasia. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPK3A
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.52 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Upk3a
Gene Accession No. O75631, Q9JKX8
Gene Alias 1110017C07Rik; proplakin IIIa; UP3a; UPIII; UPIIIA; Upk3; UPK3A; uroplakin 3; uroplakin 3A; uroplakin III; Uroplakin-3a
Gene Symbols Upk3a
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22270, 315190, 7380
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.