Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UPK3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552696
Description
Immunogen sequence: ELVPYTPQIT AWDLEGKVTA TTFSLEQPRC VFDGLASASD TVWLVVA Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 81%.
Component of the asymmetric unit membrane (AUM); a highly specialized biomembrane elaborated by terminally differentiated urothelial cells. May play an important role in AUM-cytoskeleton interaction in terminally differentiated urothelial cells. It also contributes to the formation of urothelial glycocalyx which may play an important role in preventing bacterial adherence.
Specifications
UPK3B | |
Polyclonal | |
Unconjugated | |
UPK3B | |
AI447729; AI957187; FLJ32198; LOC105375355; MGC10902; p35; PMS2L14; UP3B; UpIIIb; UPK3B; uroplakin 3B; uroplakin IIIb; Uroplakin-3b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
105375355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BT76 | |
UPK3B | |
Recombinant protein corresponding to Human UPK3B. Recombinant protein control fragment (Product #RP-90239). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction