Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UPK3BL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571312
Description
This target displays homology in the following species: Horse: 83%; Human: 100%; Pig: 83%.
UPK3BL1 is a protein coding gene.
Specifications
UPK3BL | |
Polyclonal | |
Unconjugated | |
UPK3BL1 | |
UPK3BL; UPK3BL1; UPK3BL2; UPLP; uroplakin 3B-like; uroplakin 3C protein; uroplakin-3b-like protein; Uroplakin-3b-like protein 1; Uroplakin-3b-like protein 2; uroplakin-3-like protein; uroplakin-like protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
100134938 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
B0FP48 | |
UPK3BL1 | |
synthetic peptide directed towards the C-terminal region of human UPK3BL. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction