Learn More
Waters Corporation UPLC Intact Mass Analysis Kit
Catalog No. 50780053 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1/Ea.
Catalog No. Quantity
50-780-053 1/Ea.
1 options
Catalog No. 50-780-053 Supplier Waters Corporation Supplier No. 176001519
UPLC Intact Mass Analysis Kit equipped with tubing and manual.
The MassPREP on-line desalting column can effectively desalt proteins prior to LC-MS analyses. Because non-volatile salts (e.g., NaCl) can suppress ionization of intact proteins leading to poor detection sensitivity, it is important to remove or significantly minimize the introduction of these compounds into the mass analyzer. The reversed-phase phenyl material contained in MassPREP on-line column successfully traps proteins, allowing the salts to be washed to waste prior to protein elution into the mass spectrometer. With an optimized LC-MS method, cycle times as low as 4 minutes (for intact antibody) and 10 minutes (for reduced antibody) are achievable.
Specifications
Quantity 1/Ea.
Product Type Column Kit