Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UPLC1/ASAP3Rabbit anti-Human, Polyclonal, Rockland™
SDP

Catalog No. 501990855
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0855 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0855 Supplier Rockland Immunochemicals Supplier No. 200401990
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This protein A purified antibody is directed against human UPLC1/ASAP3 protein.A BLAST analysis was used to suggest cross-reactivity with UPLC1/ASAP3 protein from mouse and rat based on approximately 80% homology with the human protein. Reactivity against homologues from other sources is not known.

This gene encodes a member of a subfamily of ADP-ribosylation factor(Arf) GTPase-activating proteins that contain additional ankyrin repeat and pleckstrin homology domains. The Arf GAP domain of this protein catalyzes the hydrolysis of GTP bound to Arf proteins. The encoded protein promotes cell differentiation and migration and has been implicated in cancer cell invasion. Alternative splicing results in multiple transcript variants.

Specifications

Antigen UPLC1/ASAP3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene Asap3
Gene Accession No. Q8TDY4
Gene Alias ACAP4; ARF6 GTPase-activating protein; Arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 3; ArfGAP with SH3 domain, ankyrin repeat and PH domain 3; ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 1; ASAP3; centaurin, beta 6; CENTB6; DDEFL1; development and differentiation enhancing factor-like 1; Development and differentiation-enhancing factor-like 1; Protein up-regulated in liver cancer 1; UPLC1; up-regulated in liver cancer 1 (UPLC1)
Gene Symbols Asap3
Host Species Rabbit
Immunogen This antibody was prepared from whole rabbit serum produced by repeated immunizations with recombinant human UPLC1/ASAP3 protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55616
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.