Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UPP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563120
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563120 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563120 Supplier Invitrogen™ Supplier No. PA563120
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGANAEKAES HNDCPVRLLN PNIAKMKEDI LYHFNLTTSR HNFP Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 63%.

UPP1 catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose- or deoxyribose-1-phosphate (PubMed:7488099). The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UPP1
Gene Accession No. Q16831
Gene Alias AI325217; im:6911242; UDRPASE; UP; UPase; UPase 1; UPP; Upp1; urdPase 1; uridine phosphorylase 1; zgc:110755
Gene Symbols UPP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UPP1. Recombinant protein control fragment (Product #RP-101551).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7378
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.