Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UPP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588945
Description
Immunogen sequence: SYTEKDKQAY LEAAYAAGVR NIEMESSVFA AMCSACGLQA AVVCVTLLNR LEGDQISSPR NVLSEYQQRP QRLVSYFIKK KLSKA; Positive Samples: HT-29, A431, A549, 293T, HeLa.
UPP1 catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose- or deoxyribose-1-phosphate (PubMed:7488099). The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis.
Specifications
UPP1 | |
Polyclonal | |
Unconjugated | |
UPP1 | |
AI325217; im:6911242; UDRPASE; UP; UPase; UPase 1; UPP; Upp1; urdPase 1; uridine phosphorylase 1; zgc:110755 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7378 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.59 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q16831 | |
UPP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 89-173 of human UPP1 (NP_0012743571). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction