Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UPP2 Monoclonal Antibody (OTI10A3), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene CF812317


Description
UPP2 Monoclonal antibody specifically detects UPP2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
UPP2 | |
Monoclonal | |
See Label | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P97576, Q99LP6, Q9HAV7 | |
Mouse | |
Protein A/G | |
RUO | |
17713, 79563, 80273 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG2b |
Western Blot | |
OTI10A3 | |
Unconjugated | |
GRPEL1 | |
1700124F02Rik; AI266885; liver-specific uridine phosphorylase; L-UrdPase; UDRPASE2; UP2; UPase 2; UPASE2; UPP2; urdPase 2; uridine phosphorylase 2; uridine phosphorylase-2 | |
Human recombinant protein fragment corresponding to amino acids 28-217 of human GRPEL1 (NP_079472) produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction