Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UPP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111057
Description
Immunogen sequence: MASVIPASNR SMRSDRNTYV GKRFVHVKNP YLDLMDEDIL YHLDLGTKTH NLPAMFGD.
Catalyzes the reversible phosphorylytic cleavage of uridine and deoxyuridine to uracil and ribose- or deoxyribose-1-phosphate (PubMed:12849978, PubMed:21855639). The produced molecules are then utilized as carbon and energy sources or in the rescue of pyrimidine bases for nucleotide synthesis (Probable). Shows broad substrate specificity and accepts uridine, deoxyuridine, and thymidine as well as the two pyrimidine nucleoside analogs 5-fluorouridine and 5-fluoro-2(')-deoxyuridine as substrates (PubMed:12849978). [UniProt].
Specifications
| UPP2 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O95045 | |
| UPP2 | |
| Recombinant Protein corresonding to Human UPP2. Recombinant protein control fragment (Product #RP-95949). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| UPP2 | |
| 1700124F02Rik; AI266885; liver-specific uridine phosphorylase; L-UrdPase; UDRPASE2; UP2; UPase 2; UPASE2; UPP2; urdPase 2; uridine phosphorylase 2; uridine phosphorylase-2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 151531 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction