Learn More
UPRT Mouse anti-Human, Clone: OTI1C9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF506869

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes uracil phosphoribosyltransferase, which catalyzes the conversion of uracil and 5-phosphoribosyl-1-R-diphosphate to uridine monophosphate (UMP). This reaction is an important part of nucleotide metabolism, specifically the pyrimidine salvage pathway. The enzyme localizes to the nucleus and cytoplasm. The protein is a potential target for rational design of drugs to treat parasitic infections and cancer.Specifications
UPRT | |
Monoclonal | |
Unconjugated | |
UPRT | |
FUR1, UPP | |
Mouse | |
Affinity Chromatography | |
RUO | |
139596 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI1C9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96BW1 | |
UPRT | |
Full length human recombit protein of human UPRT produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.