Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UPRT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551456
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551456 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551456 Supplier Invitrogen™ Supplier No. PA551456
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TGYKYEGVKF EKGNCGVSIM RSGEAMEQGL RDCCRSIRIG KILIQSDEET QRAKVYYAKF PPDIYRRKVL LMYPILSTGN TVIEAVKVLI EHGVQPSVII LLSLFSTPHG AKSIIQEFPE IT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene encodes uracil phosphoribosyltransferase, which catalyzes the conversion of uracil and 5-phosphoribosyl-1-R-diphosphate to uridine monophosphate (UMP). This reaction is an important part of nucleotide metabolism, specifically the pyrimidine salvage pathway. The enzyme localizes to the nucleus and cytoplasm. The protein is a potential target for rational design of drugs to treat parasitic infections and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UPRT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UPRT
Gene Accession No. Q96BW1
Gene Alias FUR1; Gm774; RP11-311P8.3; UMP pyrophosphorylase; UPP; Uprt; UPRTase; uracil phosphoribosyltransferase (FUR1) homolog; uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae); uracil phosphoribosyltransferase homolog
Gene Symbols UPRT
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UPRT. Recombinant protein control fragment (Product #RP-91918).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139596
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.