Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Upstream Binding Protein 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987454
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7454 Supplier Bioss Supplier No. BS6730R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBP1 functions as a transcriptional activator in a promoter context-dependent manner. Modulates the placental expression of CYP11A1. Involved in regulation of the alpha-globin gene in erythroid cells. Activation of the alpha-globin promoter in erythroid cells is via synergistic interaction with TFCP2. Involved in regulation of the alpha-globin gene in erythroid cells. Binds strongly to sequences around the HIV-1 initiation site and weakly over the TATA-box. Represses HIV-1 transcription by inhibiting the binding of TFIID to the TATA-box.

Specifications

Antigen Upstream Binding Protein 1
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene UBP1
Gene Accession No. Q9NZI7
Gene Alias Cp2b; LBP1; LBP1A; LBP-1a; LBP1B; LBP-1b; Nf2d9; Nuclear factor 2d9; RGD1564700; Transcription factor LBP-1; UBP1; upstream binding protein 1; upstream binding protein 1 (LBP-1a); Upstream-binding protein 1
Gene Symbols UBP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Upstream Binding Protein 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7342
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.