Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114251
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114251 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114251 Supplier Invitrogen™ Supplier No. PA5114251
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RCQMPDTFNS WFLITLLHVW MCLVRMKQEG RSGKYMCRII VHFMWEDVQQ For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCC1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene UQCC1
Gene Accession No. Q9NVA1
Gene Alias 2310079L17Rik; 2410003P15Rik; 3110038N19Rik; basic FGF repressed, Zic binding protein; basic FGF repressed, zinc binding protein; basic FGF-repressed Zic-binding protein; bFGF-repressed Zic-binding protein; BFZB; Bfzp; BZFB; C20orf44; Cbp3; cytochrome B protein synthesis 3 homolog; FLJ10850; mbFZb; ubiquinol-cytochrome c reductase complex assembly factor 1; ubiquinol-cytochrome c reductase complex chaperone; ubiquinol-cytochrome c reductase complex chaperone CBP3 homolog; ubiquinol-cytochrome c reductase complex chaperone, CBP3 homolog; ubiquinol-cytochrome-c reductase complex assembly factor 1; UQCC; UQCC1
Gene Symbols UQCC1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Human UQCC (aa 136-185).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55245
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.