Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UQCC1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501570407
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0407 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0407 Supplier Bethyl Laboratories, Inc Supplier No. A305430AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family.

Specifications

Antigen UQCC1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene UQCC1
Gene Accession No. Q9CWU6, Q9NVA1
Gene Alias Basic FGF-repressed Zic-binding protein, bFGF-repressed Zic-binding protein, BFZB, C20orf44, CBP3, chromosome 20 open reading frame 44, cytochrome B protein synthesis 3 homolog, cytochrome B protein synthesis 3 homolog (S. cerevisiae), FLJ10850, ubiquinol-cytochrome c reductase complex chaperone, ubiquinol-cytochrome c reductase complex chaperone CBP3 homolog, UQCC
Gene Symbols Uqcc1
Host Species Rabbit
Immunogen Between 249 and 299
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55245, 56046
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.