Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106324
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106324 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106324 Supplier Invitrogen™ Supplier No. PA5106324
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UQCRB.

UQCRB encodes a product that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This component is involved in redox-linked proton pumping.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene UQCRB
Gene Accession No. P14927, Q9D855
Gene Alias 2210415M14Rik; Complex III subunit 7; complex III subunit VII; Cytochrome b-c1 complex subunit 7; MC3DN3; mitochondrial ubiquinone-binding protein; QCR7; QPC; QP-C; ubiquinol-cytochrome c reductase binding protein; ubiquinol-cytochrome c reductase binding protein-like; ubiquinol-cytochrome c reductase complex 14 kDa protein; ubiquinol-cytochrome c reductase, complex III subunit VI; UQBC; UQBP; UQCR6; Uqcrb; Uqcrbl; UQPC
Gene Symbols UQCRB
Host Species Rabbit
Immunogen A synthesized peptide derived from human UQCRB(Accession P14927), corresponding to amino acid residues M1-L51.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362897, 67530, 7381
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.