Learn More
Invitrogen™ UQCRB Polyclonal Antibody

Description
UQCRB Polyclonal Antibody for Western Blot, IHC (P), IP
Specifications
Specifications
| Antigen | UQCRB |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.03% ProClin 300; pH 7 |
| Gene | UQCRB |
| Gene Accession No. | P14927 |
| Gene Alias | 2210415M14Rik; Complex III subunit 7; complex III subunit VII; Cytochrome b-c1 complex subunit 7; MC3DN3; mitochondrial ubiquinone-binding protein; QCR7; QPC; QP-C; ubiquinol-cytochrome c reductase binding protein; ubiquinol-cytochrome c reductase binding protein-like; ubiquinol-cytochrome c reductase complex 14 kDa protein; ubiquinol-cytochrome c reductase, complex III subunit VI; UQBC; UQBP; UQCR6; Uqcrb; Uqcrbl; UQPC |
| Gene Symbols | UQCRB |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.