Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UQCRC1 Antibody (OTI1G6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274796PE
Description
UQCRC1 Monoclonal specifically detects UQCRC1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Flow Cytometry.Specifications
UQCRC1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
UQCRC1 | |
Recombinant protein fragment corresponding to amino acids 46-305 of human UQCRC1 (NP_003356) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Primate, Monkey | |
IgG1 |
Flow Cytometry | |
OTI1G6 | |
Flow Cytometry | |
Complex III subunit 1, Core protein I, cytochrome b-c1 complex subunit 1, mitochondrial, D3S3191, QCR1, ubiquinol-cytochrome c reductase core protein I, Ubiquinol-cytochrome-c reductase complex core protein 1, UQCR1 | |
Mouse | |
Affinity Purified | |
metabolism | |
7384.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction