Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Complex III Monoclonal Antibody (11A51H12)
GREENER_CHOICE

Catalog No. 439400
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-940-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-940-0 Supplier Invitrogen™ Supplier No. 439400
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: Bovine heart and Human mitochondria.

Complex III, or ubiquinol cytochrome c oxidoreductase, is a complex of 11 subunits and catalyses electron transfer from reduced ubiquinone or coenzyme Q 10 to cytochrome c. The complex contains 11 different subunits, one of which, cytochrome b, is encoded on mitochondrial DNA, while the remainder subunits are nuclear-encoded. Genetic complex III deficiencies occur but are rare.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Complex III
Applications Immunoprecipitation
Classification Monoclonal
Clone 11A51H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation HEPES buffered saline with 0.02% sodium azide
Gene UQCRQ
Gene Accession No. O14949, O14957, P00125, P00126, P00129, P00130, P00156, P07552, P07919, P08574, P13271, P13272, P14927, P22695, P23004, P31800, P31930, P47985, Q9UDW1
Gene Alias 0710008D09Rik; 1100001F06Rik; 1110020P15Rik; 1110032G10Rik; 1500004O06Rik; 1500040F11Rik; 2210415M14Rik; 2210416J04Rik; 2310021J10Rik; 2610002H19Rik; 2610041P16Rik; 4430402G14Rik; 5830407L17Rik; 8 kDa subunit 9; 9.5kDa; AA408921; AA959903; AA960494; AI875505; AL022707; c1502; COB; Complex III subunit 1; complex III subunit 10; Complex III subunit 2; Complex III subunit 3; Complex III subunit 4; complex III subunit 5; Complex III subunit 6; complex III subunit 7; Complex III subunit 8; complex III subunit 9; Complex III subunit III; Complex III subunit IV; Complex III subunit IX; Complex III subunit VII; complex III subunit VIII; Complex III subunit X; complex III subunit XI; Core protein I; core protein II; CYC1; CYTB; Cytochrome b; cytochrome bc-1 complex core protein II; Cytochrome b-c1 complex subunit 1, mitochondrial; cytochrome b-c1 complex subunit 10; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 2, mitochondrial; Cytochrome b-c1 complex subunit 3; Cytochrome b-c1 complex subunit 4; cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 6, mitochondrial; cytochrome b-c1 complex subunit 7; Cytochrome b-c1 complex subunit 8; cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; cytochrome c1; cytochrome c-1; cytochrome c1 non-heme 11 kDa protein; Cytochrome c1 non-heme 7 kDa protein; cytochrome c1, heme protein, mitochondrial; cytochrome C1, nonheme 11 kDa protein; cytochrome C1, nonheme 7kDa protein; D3S3191; HSPC051; HSPC119; HSPC151; hypothetical protein LOC613899; hypothetical protein LOC616871; liver regeneration-related protein LRRGT00195; Low molecular mass ubiquinone-binding protein; low molecular mass ubiquinone-binding protein (9.5kD); low molecular mass ubiquinone-binding protein (Qpc-pending); LRRGT00195; MC3DN3; MC3DN4; MC3DN5; MC3DN6; Mitochondrial hinge protein; mitochondrial ubiquinone-binding protein; mitochondrially encoded cytochrome b; MTCYB; MT-CYB; QCR1; QCR10; QCR2; QCR6; QCR7; QCR8; QCR9; QPC; QP-C; Rieske iron-sulfur; Rieske iron-sulfur protein; Rieske iron-sulfur protein precursor; Rieske protein UQCRFS1; RIP1; RIS1; RISP; similar to ubiquinol-cytrochrome-c reductase (subunit II); smallest subunit of ubiquinol-cytochrome c reductase; Su9; ubiquinol cytochrome c reductase core protein 2; ubiquinol cytochrome c reductase hinge protein; ubiquinol cytochrome c reductase, 6.4kDa subunit; ubiquinol--cytochrome c reductase; ubiquinol-cytochrome c reductase (6.4kD) subunit; Ubiquinol-cytochrome c reductase 8 kDa protein; ubiquinol-cytochrome c reductase binding protein; ubiquinol-cytochrome c reductase binding protein-like; ubiquinol-cytochrome c reductase complex (7.2 kD); ubiquinol-cytochrome c reductase complex 11 kDa protein; Ubiquinol-cytochrome c reductase complex 14 kDa protein; ubiquinol-cytochrome c reductase complex 6.4 kDa protein; Ubiquinol-cytochrome c reductase complex 7.2 kDa protein; ubiquinol-cytochrome c reductase complex 7.2kDa protein; ubiquinol-cytochrome c reductase complex 9.5 kDa protein; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase complex III subunit VII; ubiquinol-cytochrome c reductase complex protein; ubiquinol-cytochrome c reductase complex ubiquinone-binding protein QP-C; ubiquinol-cytochrome c reductase core protein 1; ubiquinol-cytochrome c reductase core protein 2; ubiquinol-cytochrome c reductase core protein I; ubiquinol-cytochrome c reductase core protein II; ubiquinol-cytochrome c reductase hinge protein; ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase subunit; ubiquinol-cytochrome c reductase, 6.4kDa subunit; ubiquinol-cytochrome c reductase, complex III subunit VI; ubiquinol-cytochrome c reductase, complex III subunit VII; ubiquinol-cytochrome c reductase, complex III subunit VII, 9.5kDa; ubiquinol-cytochrome c reductase, complex III subunit VIII
Gene Symbols CYC1, MT-CYB, UQCR10, UQCR11, UQCRB, UQCRC1, UQCRC2, UQCRFS1, UQCRH, UQCRQ
Host Species Mouse
Immunogen Bovine Heart Complex III.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10975, 1537, 27089, 281570, 282393, 282394, 286885, 287020, 29796, 4519, 512500, 613899, 616109, 616871, 7381, 7384, 7385, 7386, 7388
Target Species Human, Bovine
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.