Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRC1 Monoclonal Antibody (GT139)
GREENER_CHOICE

Catalog No. PIMA527790
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527790 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527790 Supplier Invitrogen™ Supplier No. MA527790
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2, mouse brain, rat brain Predicted Reactivity: Rhesus Monkey (97%), Chimpanzee (100%), Bovine (88%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

UQCRC1 encodes a product that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRC1
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT139
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Uqcrc1
Gene Accession No. P31930, Q68FY0, Q9CZ13
Gene Alias 1110032G10Rik; Complex III subunit 1; core protein I; Cytochrome b-c1 complex subunit 1, mitochondrial; D3S3191; hypothetical protein MGC73404; QCR1; ubiquinol--cytochrome c reductase; ubiquinol-cytochrome c reductase core I protein; ubiquinol-cytochrome c reductase core protein 1; ubiquinol-cytochrome c reductase core protein I; ubiquinol-cytochrome-c reductase complex core protein 1; UQCR1; Uqcrc1; UQCRC1 protein; zgc:73404; zgc:85750
Gene Symbols Uqcrc1
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human UQCRC1. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22273, 301011, 7384
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.