Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100121 Supplier Invitrogen™ Supplier No. PA5100121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UQCRC1.

UQCRC1 encodes a product that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Uqcrc1
Gene Accession No. P31930, Q68FY0, Q9CZ13
Gene Alias 1110032G10Rik; Complex III subunit 1; core protein I; Cytochrome b-c1 complex subunit 1, mitochondrial; D3S3191; hypothetical protein MGC73404; QCR1; ubiquinol--cytochrome c reductase; ubiquinol-cytochrome c reductase core I protein; ubiquinol-cytochrome c reductase core protein 1; ubiquinol-cytochrome c reductase core protein I; ubiquinol-cytochrome-c reductase complex core protein 1; UQCR1; Uqcrc1; UQCRC1 protein; zgc:73404; zgc:85750
Gene Symbols Uqcrc1
Host Species Rabbit
Immunogen A synthesized peptide derived from human UQCRC1(Accession P31930), corresponding to amino acid residues G391-V441.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22273, 301011, 7384
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.