Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582101 Supplier Invitrogen™ Supplier No. PA582101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDALPFAHVA IAVEGPGWAS PDNVALQVAN AIIGHYDCTY GGGVHLSSPL ASGAVANKLC QSFQTFSICY AETGLLGAHF VCDRMKIDDM MFVLQGQWMR LCTSATESEV ARGKNILRNA LVSHLDGTTP VCEDIGR Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 91%.

UQCRC1 encodes a product that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Uqcrc1
Gene Accession No. P31930
Gene Alias 1110032G10Rik; Complex III subunit 1; core protein I; Cytochrome b-c1 complex subunit 1, mitochondrial; D3S3191; hypothetical protein MGC73404; QCR1; ubiquinol--cytochrome c reductase; ubiquinol-cytochrome c reductase core I protein; ubiquinol-cytochrome c reductase core protein 1; ubiquinol-cytochrome c reductase core protein I; ubiquinol-cytochrome-c reductase complex core protein 1; UQCR1; Uqcrc1; UQCRC1 protein; zgc:73404; zgc:85750
Gene Symbols Uqcrc1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UQCRC1. Recombinant protein control fragment (Product #RP-102222).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7384
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.