Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UQCRC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598231
Description
UQCRC1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
UQCRC1 encodes a product that is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.
Specifications
UQCRC1 | |
Polyclonal | |
Unconjugated | |
Uqcrc1 | |
1110032G10Rik; Complex III subunit 1; core protein I; Cytochrome b-c1 complex subunit 1, mitochondrial; D3S3191; hypothetical protein MGC73404; QCR1; ubiquinol--cytochrome c reductase; ubiquinol-cytochrome c reductase core I protein; ubiquinol-cytochrome c reductase core protein 1; ubiquinol-cytochrome c reductase core protein I; ubiquinol-cytochrome-c reductase complex core protein 1; UQCR1; Uqcrc1; UQCRC1 protein; zgc:73404; zgc:85750 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22273, 7384 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.68 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P31930, Q9CZ13 | |
Uqcrc1 | |
Recombinant Human Cytochrome b-c1 complex subunit 1, mitochondrial protein (156-480AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction