Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UQCRC2 Antibody (2F6), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP315182
This item is not returnable.
View return policy
Description
UQCRC2 Monoclonal antibody specifically detects UQCRC2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
UQCRC2 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.4), 150mM NaCl, and 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry | |
2F6 | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
Complex III subunit 2, Core protein II, cytochrome b-c1 complex subunit 2, mitochondrial, QCR2, ubiquinol-cytochrome c reductase core protein II, Ubiquinol-cytochrome-c reductase complex core protein 2, UQCR2 | |
A synthesized peptide derived from human UQCRC2. UniProt ID (P22695). | |
100 μg | |
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
7385 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction