Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UQCRC2 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297364IR
Description
UQCRC2 Polyclonal specifically detects UQCRC2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
UQCRC2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
7385 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 755 | |
Complex III subunit 2, Core protein II, cytochrome b-c1 complex subunit 2, mitochondrial, QCR2, ubiquinol-cytochrome c reductase core protein II, Ubiquinol-cytochrome-c reductase complex core protein 2, UQCR2 | |
Produced in rabbits immunized with E. coli-derived Human UQCRC2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction