Learn More
Invitrogen™ UQCRC2 Polyclonal Antibody

Description
Positive Control: Rat heart tissue, human liver tissue, mouse brain tissue, HepG2. Subcellular Location: Mitochondrion.
Specifications
Specifications
| Antigen | UQCRC2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | UQCRC2 |
| Gene Accession No. | P22695, P32551, Q9DB77 |
| Gene Alias | 1500004O06Rik; complex III subunit 2; Core protein II; cytochrome bc-1 complex core protein II; cytochrome b-c1 complex subunit 2, mitochondrial; MC3DN5; QCR2; U MC3DN5; ubiquinol cytochrome c reductase core protein 2; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase core protein II; ubiquinol-cytochrome-c reductase complex core protein 2; UQCR2; Uqcrc2 |
| Gene Symbols | UQCRC2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.