Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530204
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530204 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530204 Supplier Invitrogen™ Supplier No. PA530204
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Jurkat, Raji, NCI-H929, mouse brain, rat brain. Predicted reactivity: Mouse (82%), Rat (81%), Rhesus Monkey (98%), Chimpanzee (99%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. The core protein 2 is required for the assembly of the complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRC2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Proximity Ligation Assay, Immunocytochemistry
Classification Polyclonal
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene UQCRC2
Gene Accession No. P22695, P32551, Q9DB77
Gene Alias 1500004O06Rik; complex III subunit 2; Core protein II; cytochrome bc-1 complex core protein II; cytochrome b-c1 complex subunit 2, mitochondrial; MC3DN5; QCR2; U MC3DN5; ubiquinol cytochrome c reductase core protein 2; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase core protein II; ubiquinol-cytochrome-c reductase complex core protein 2; UQCR2; Uqcrc2
Gene Symbols UQCRC2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human UQCRC2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293448, 67003, 7385
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.