Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRFS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106341
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106341 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106341 Supplier Invitrogen™ Supplier No. PA5106341
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total UQCRFS1.

Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRFS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Uqcrfs1
Gene Accession No. P20788, P47985, Q9CR68
Gene Alias 4430402G14Rik; 8 kDa subunit 9; AI875505; Complex III subunit 5; Complex III subunit IX; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; liver regeneration-related protein LRRGT00195; LRRGT00195; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RIP1; RIS1; RISP; Su9; Ubiquinol-cytochrome c reductase 8 kDa protein; Ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1; UQCR5; UQCRFS1
Gene Symbols Uqcrfs1
Host Species Rabbit
Immunogen A synthesized peptide derived from human UQCRFS1(Accession P47985), corresponding to amino acid residues V205-P255.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291103, 66694, 7386
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.