Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
UQCRH Antibody (SR1085), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32250850UL
Description
UQCRH Monoclonal antibody specifically detects UQCRH in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
UQCRH | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR1085 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Complex III subunit 6, Complex III subunit VIII, cytochrome b-c1 complex subunit 6, mitochondrial, Cytochrome c1 non-heme 11 kDa protein, MGC111572, Mitochondrial hinge protein, QCR6, Ubiquinol-cytochrome c reductase complex 11 kDa protein, ubiquinol-cytochrome c reductase hinge protein, ubiquinol-cytochrome c reductase, complex III subunit VIII, UQCR8 | |
A synthesized peptide derived from human UQCRH (Uniprot #: P07919) | |
50 μg | |
Core ESC Like Genes, Stem Cell Markers | |
7388 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction