Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UQCRH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560030
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560030 Supplier Invitrogen™ Supplier No. PA560030
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TTVREQCEQL EKCVKARERL ELCDERVSSR SHTEEDCTEE LFDFLHARDH CVAHKLFNNL K Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.

Cytochrome b-c1 complex subunit 6, mitochondrial is a protein that in humans is encoded by the UQCRHgene. This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UQCRH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene UQCRH
Gene Accession No. P07919
Gene Alias 2210416J04Rik; 2310021J10Rik; 2610041P16Rik; Complex III subunit 6; complex III subunit VIII; Cytochrome b-c1 complex subunit 6, mitochondrial; cytochrome c1 non-heme 11 kDa protein; cytochrome C1, nonheme 11 kDa protein; mitochondrial hinge protein; QCR6; ubiquinol cytochrome c reductase hinge protein; ubiquinol-cytochrome c reductase complex 11 kDa protein; ubiquinol-cytochrome c reductase hinge protein; ubiquinol-cytochrome c reductase, complex III subunit VIII; UQCR8; UQCRH
Gene Symbols UQCRH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human UQCRH. Recombinant protein control fragment (Product #RP-99817).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7388
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.