Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UQCRH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501727927 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7927 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7927 Supplier Proteintech Group Inc Supplier No. 123641AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cytochrome b-c1 complex subunit 6, mitochondrial is a protein that in humans is encoded by the UQCRHgene. This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. This protein may mediate formation of the complex between cytochromes c and c1.

Specifications

Antigen UQCRH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene UQCRH
Gene Accession No. P07919, P99028, Q5M9I5
Gene Alias Complex III subunit 6, Complex III subunit VIII, Mitochondrial hinge protein, QCR6, UQCRH
Gene Symbols Uqcrh
Host Species Rabbit
Immunogen UQCRH Fusion Protein Ag3037
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366448, 66576, 7388
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.